Product Details

SNP ID
rs7176592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40384796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGATAAACAAGTTTCCTTCCCTT[C/T]CTTGACACCAGCCATCCAGTTCCTC
Phenotype
MIM: 607503 MIM: 614718
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DISP2 PubMed Links

Gene Details

Gene
DISP2
Gene Name
dispatched RND transporter family member 2
There are no transcripts associated with this gene.

Gene
KNSTRN
Gene Name
kinetochore localized astrin/SPAG5 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142761.1 Intron NP_001136233.1
NM_001142762.1 Intron NP_001136234.1
NM_033286.3 Intron NP_150628.3

View Full Product Details