Product Details

SNP ID
rs4923867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40444828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCAGATAGTGAAAAAATATATT[C/T]TGCATCCATCCCCTTTCCCCCAAAC
Phenotype
MIM: 613880 MIM: 607036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAHD1 PubMed Links

Gene Details

Gene
BAHD1
Gene Name
bromo adjacent homology domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301132.1 Intron NP_001288061.1
NM_014952.4 Intron NP_055767.3
Gene
IVD
Gene Name
isovaleryl-CoA dehydrogenase
There are no transcripts associated with this gene.

View Full Product Details