Product Details

SNP ID
rs7250536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44504582 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTGCTTTCCTTCTAGGAGTCTGG[A/C]GCTTCAGTGACTGACCCTTTATAAA
Phenotype
MIM: 606740
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM20 PubMed Links

Gene Details

Gene
CEACAM20
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102597.2 4040 Intron NP_001096067.2
NM_001102598.2 4040 Intron NP_001096068.2
NM_001102599.2 4040 Intron NP_001096069.2
NM_001102600.2 4040 Intron NP_001096070.2
XM_011526429.2 4040 UTR 3 XP_011524731.1
XM_011526430.2 4040 UTR 3 XP_011524732.1
XM_011526431.2 4040 Intron XP_011524733.1
XM_011526432.2 4040 UTR 3 XP_011524734.1
Gene
ZNF180
Gene Name
zinc finger protein 180
There are no transcripts associated with this gene.

View Full Product Details