Product Details

SNP ID
rs6510515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36055325 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACGGCCCCTCCCACTTTCTCCT[C/G]GCTGCTTTCCCGGCTCCACCCTCAG
Phenotype
MIM: 612536 MIM: 613583
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927572 PubMed Links

Gene Details

Gene
LOC101927572
Gene Name
uncharacterized LOC101927572
There are no transcripts associated with this gene.

Gene
THAP8
Gene Name
THAP domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152658.2 Intron NP_689871.1
XM_005258623.2 Intron XP_005258680.1
XM_011526604.2 Intron XP_011524906.1
Gene
WDR62
Gene Name
WD repeat domain 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083961.1 Intron NP_001077430.1
NM_173636.4 Intron NP_775907.4
XM_005258809.2 Intron XP_005258866.1
XM_011526837.1 Intron XP_011525139.1
XM_011526838.1 Intron XP_011525140.1
XM_011526839.1 Intron XP_011525141.1
XM_011526840.2 Intron XP_011525142.1
XM_011526841.2 Intron XP_011525143.1
XM_011526842.1 Intron XP_011525144.1
XM_011526843.1 Intron XP_011525145.1
XM_011526844.2 Intron XP_011525146.1
XM_017026665.1 Intron XP_016882154.1

View Full Product Details