Product Details
- SNP ID
-
rs6661046
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:24416733 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTCACCAGGTGAAAAGTATGTGTC[G/T]TATAAGGGAGTACAGCTTGCAAAGG
- Phenotype
-
MIM: 615826
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NIPAL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115885613] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NIPAL3
- Gene Name
- NIPA like domain containing 3
- Gene
- STPG1
- Gene Name
- sperm tail PG-rich repeat containing 1
There are no transcripts associated with this gene.
View Full Product Details