Product Details

SNP ID
rs7778391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150518861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATCTATTTGGAATTTGTATGA[C/T]ATGAGGAATTGGAGGCAATTTTATC
Phenotype
MIM: 616961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIMAP7 PubMed Links

Gene Details

Gene
GIMAP7
Gene Name
GTPase, IMAP family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153236.3 Intron NP_694968.1

View Full Product Details