Product Details

SNP ID
rs6961774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150453513 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCTCTCCCCTGAGTGCCACAGAC[C/T]CCATTTTTTGAAAAGGTATGAACCA
Phenotype
MIM: 616962
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIMAP8 PubMed Links

Gene Details

Gene
GIMAP8
Gene Name
GTPase, IMAP family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175571.3 Intron NP_783161.1
XM_005249950.4 Intron XP_005250007.1
XM_005249951.3 Intron XP_005250008.1
Gene
LINC00996
Gene Name
long intergenic non-protein coding RNA 996
There are no transcripts associated with this gene.

View Full Product Details