Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_145292.3 | 896 | Intron | NP_660335.2 | ||
XM_006715862.3 | 896 | Intron | XP_006715925.1 | ||
XM_011515849.2 | 896 | Intron | XP_011514151.1 | ||
XM_011515852.2 | 896 | Intron | XP_011514154.1 | ||
XM_011515853.2 | 896 | Intron | XP_011514155.1 | ||
XM_017011793.1 | 896 | UTR 5 | XP_016867282.1 | ||
XM_017011794.1 | 896 | Intron | XP_016867283.1 | ||
XM_017011795.1 | 896 | UTR 5 | XP_016867284.1 | ||
XM_017011796.1 | 896 | UTR 5 | XP_016867285.1 |