Product Details

SNP ID
rs6970768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151148971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCTCCAGTCCCCAGGCTTCCC[C/T]TTAGCTTCGGTTTAGGTCCCAGAAG
Phenotype
MIM: 616813 MIM: 603354
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGAP3 PubMed Links

Gene Details

Gene
AGAP3
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
There are no transcripts associated with this gene.

Gene
GBX1
Gene Name
gastrulation brain homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098834.2 724 Missense Mutation AAG,AGG K237R NP_001092304.1
XM_017011964.1 724 Missense Mutation AAG,AGG K78R XP_016867453.1
XM_017011965.1 724 Missense Mutation AAG,AGG K77R XP_016867454.1

View Full Product Details