Product Details

SNP ID
rs7212854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44177049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCGCTGCACAACGCTTGTGCCA[A/G]CGGCTGCGGGGGCCTGGCCGAGCTG
Phenotype
MIM: 615056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB16 PubMed Links

Gene Details

Gene
ASB16
Gene Name
ankyrin repeat and SOCS box containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080863.4 965 Missense Mutation AAC,AGC N294S NP_543139.4
Gene
ASB16-AS1
Gene Name
ASB16 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TMUB2
Gene Name
transmembrane and ubiquitin like domain containing 2
There are no transcripts associated with this gene.

View Full Product Details