Product Details

SNP ID
rs7057844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103356540 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTTCCCGTCTGGATACCAGCTT[C/T]CTTCAGCAGCGCAGGCGGTGGTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCEAL9 PubMed Links

Gene Details

Gene
TCEAL9
Gene Name
transcription elongation factor A like 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006612.1 89 UTR 5 NP_001006613.1
NM_001006613.1 89 UTR 5 NP_001006614.1
NM_001006614.1 89 UTR 5 NP_001006615.1
NM_016303.2 89 UTR 5 NP_057387.1

View Full Product Details