Product Details
- SNP ID
-
rs4349869
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:64069578 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCAAAGAAAAGAGCCTTGGAATGC[A/G]AGGAGACAGAAGACAGTAGCCAAAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF727
PubMed Links
Gene Details
- Gene
- ZNF727
- Gene Name
- zinc finger protein 727
View Full Product Details