Product Details

SNP ID
rs6677424
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:147636115 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTTGGTCTCCCAGAGTGTAGCT[A/G]TGGTAAAGCAGAAGACAGAGAAACC
Phenotype
MIM: 611471 MIM: 602597
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACP6 PubMed Links

Gene Details

Gene
ACP6
Gene Name
acid phosphatase 6, lysophosphatidic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323625.1 Intron NP_001310554.1
NM_016361.4 Intron NP_057445.4
XM_011509601.2 Intron XP_011507903.1
XM_017001422.1 Intron XP_016856911.1
Gene
BCL9
Gene Name
B-cell CLL/lymphoma 9
There are no transcripts associated with this gene.

View Full Product Details