Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022336.3 | 1679 | UTR 3 | NP_071731.1 | ||
XM_006712204.1 | 1679 | UTR 3 | XP_006712267.1 | ||
XM_011510502.2 | 1679 | UTR 3 | XP_011508804.2 | ||
XM_011510503.2 | 1679 | UTR 3 | XP_011508805.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_006267.4 | 1679 | Intron | NP_006258.3 | ||
XM_005264002.2 | 1679 | Intron | XP_005264059.1 | ||
XM_005264003.2 | 1679 | Intron | XP_005264060.1 | ||
XM_005264004.2 | 1679 | Intron | XP_005264061.1 | ||
XM_005264005.4 | 1679 | Intron | XP_005264062.1 | ||
XM_005264007.2 | 1679 | Intron | XP_005264064.1 | ||
XM_011511575.2 | 1679 | Intron | XP_011509877.1 | ||
XM_011511576.2 | 1679 | Intron | XP_011509878.1 | ||
XM_011511578.2 | 1679 | Intron | XP_011509880.1 | ||
XM_017004623.1 | 1679 | Intron | XP_016860112.1 | ||
XM_017004624.1 | 1679 | Intron | XP_016860113.1 | ||
XM_017004625.1 | 1679 | Intron | XP_016860114.1 |