Product Details
- SNP ID
-
rs5752326
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:26448024 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGAAACAGGCTCAGAGCGGGCAG[C/T]TGACATGAACAAGGTCACGCAGCCA
- Phenotype
-
MIM: 606682
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ASPHD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112443942] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ASPHD2
- Gene Name
- aspartate beta-hydroxylase domain containing 2
There are no transcripts associated with this gene.
- Gene
- HPS4
- Gene Name
- HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
View Full Product Details