Product Details
- SNP ID
-
rs8086898
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:63908200 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGAATTTGTCTGTATAAGTCAGA[C/G]AGTAACCTGAGCTGGTTTGAAGTTT
- Phenotype
-
MIM: 602058
MIM: 173390
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SERPINB10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114997242] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SERPINB10
- Gene Name
- serpin family B member 10
- Gene
- SERPINB2
- Gene Name
- serpin family B member 2
There are no transcripts associated with this gene.
View Full Product Details