Product Details

SNP ID
rs7942897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61521350 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGCTCTAATTGGTATCTAAGGAA[C/T]ATCCGACACCAAGAGGCGAAGCTTC
Phenotype
MIM: 604146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYT7 PubMed Links

Gene Details

Gene
SYT7
Gene Name
synaptotagmin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252065.1 Intron NP_001238994.1
NM_001300773.1 Intron NP_001287702.1
NM_004200.3 Intron NP_004191.2
XM_005274383.4 Intron XP_005274440.1
XM_005274384.2 Intron XP_005274441.1
XM_005274385.4 Intron XP_005274442.1
XM_005274387.4 Intron XP_005274444.1
XM_005274390.4 Intron XP_005274447.1
XM_006718736.3 Intron XP_006718799.1
XM_011545335.2 Intron XP_011543637.1
XM_011545336.2 Intron XP_011543638.1
XM_011545337.2 Intron XP_011543639.1
XM_011545338.2 Intron XP_011543640.1
XM_011545339.2 Intron XP_011543641.1
XM_011545340.2 Intron XP_011543642.1
XM_011545341.2 Intron XP_011543643.1
XM_011545342.2 Intron XP_011543644.1
XM_011545343.2 Intron XP_011543645.1

View Full Product Details