Product Details

SNP ID
rs7160183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:66497474 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTATCATATTTAGCAAATAAAAA[A/G]GGATACCCTATTAAATTTGAATTTC
Phenotype
MIM: 603930
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPHN PubMed Links

Gene Details

Gene
GPHN
Gene Name
gephyrin
There are no transcripts associated with this gene.

Gene
LINC00238
Gene Name
long intergenic non-protein coding RNA 238
There are no transcripts associated with this gene.

View Full Product Details