Product Details

SNP ID
rs8061916
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46473222 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTAAGGCTACCATTTTCTGGCCA[C/G]CCTTCCTACAGTCACATAGCTTTGT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ANKRD26P1 PubMed Links

Gene Details

Gene
ANKRD26P1
Gene Name
ankyrin repeat domain 26 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details