Product Details

SNP ID
rs7192932
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46898215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAGCGCCGCCTCGGGAGATTGC[C/T]GGGCCTTGGTGAGGCGATGAGTGCA
Phenotype
MIM: 138210
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GPT2 PubMed Links

Gene Details

Gene
GPT2
Gene Name
glutamic--pyruvic transaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142466.2 Intron NP_001135938.1
NM_133443.3 Intron NP_597700.1
XM_017023790.1 Intron XP_016879279.1

View Full Product Details