Product Details

SNP ID
rs6975566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141652515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATATTTGAAAATTATAACCATGTT[G/T]TTCAAATCCCAAAATACTCTGGGAG
Phenotype
MIM: 610345
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGK PubMed Links

Gene Details

Gene
AGK
Gene Name
acylglycerol kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018238.3 Intron NP_060708.1
XM_005250023.3 Intron XP_005250080.1
XM_011516397.2 Intron XP_011514699.1
Gene
KIAA1147
Gene Name
KIAA1147
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080392.1 Intron NP_001073861.1

View Full Product Details