Product Details

SNP ID
rs7068341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16592300 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACATACCATTTTTTTTGTGTCGGC[C/T]TTCTGGGGTTGACAGCCCTTTCAGT
Phenotype
MIM: 179555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSU1 PubMed Links

Gene Details

Gene
RSU1
Gene Name
Ras suppressor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012425.3 2071 UTR 3 NP_036557.1
NM_152724.2 2071 UTR 3 NP_689937.2
XM_005252552.3 2071 UTR 3 XP_005252609.1

View Full Product Details