Product Details

SNP ID
rs7831137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:74988277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATCGTTATTTAAAATGTATTT[A/C]TTCTGAGAAAACAAACCTTTAAACT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRISPLD1 PubMed Links

Gene Details

Gene
CRISPLD1
Gene Name
cysteine rich secretory protein LCCL domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286777.1 Intron NP_001273706.1
NM_001286778.1 Intron NP_001273707.1
NM_031461.5 Intron NP_113649.1

View Full Product Details