Product Details
- SNP ID
-
rs6565375
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:29306540 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAGCTGCGCTTCCCAGGATGTTCA[T/A]GGAGTGACCGTGGAGCTGCGCCTCC
- Phenotype
-
- Polymorphism
- T/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SNX29P2
PubMed Links
Gene Details
- Gene
- SNX29P2
- Gene Name
- sorting nexin 29 pseudogene 2
There are no transcripts associated with this gene.
View Full Product Details