Product Details

SNP ID
rs4962140
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133407383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTGCAGGGTGATGGCAGGTTGGG[C/T]GCTGGGCACAGGGGCACAAGGACCA
Phenotype
MIM: 604134 MIM: 602930
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADAMTS13 PubMed Links

Gene Details

Gene
ADAMTS13
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 13
There are no transcripts associated with this gene.

Gene
REXO4
Gene Name
REX4 homolog, 3'-5' exonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001279349.1 Intron NP_001266278.1
NM_001279350.1 Intron NP_001266279.1
NM_001279351.1 Intron NP_001266280.1
NM_020385.3 Intron NP_065118.2
Gene
STKLD1
Gene Name
serine/threonine kinase like domain containing 1
There are no transcripts associated with this gene.

View Full Product Details