Product Details
- SNP ID
-
rs6955800
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:8438750 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCAATGCCCCACACCAAGCCACAG[G/T]TTCCTACCTGTATTTGGGTGAATGT
- Phenotype
-
MIM: 604639
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NXPH1
PubMed Links
Gene Details
- Gene
- NXPH1
- Gene Name
- neurexophilin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152745.2 |
|
Intron |
|
|
NP_689958.1 |
View Full Product Details