Product Details
- SNP ID
-
rs6875694
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:72111160 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTAGCTAGTGGTTCTCATTTTCA[G/T]CAAAACCCTGCTTCTTGGTTTTATT
- Phenotype
-
MIM: 157129
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MAP1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111406578] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAP1B
- Gene Name
- microtubule associated protein 1B
View Full Product Details