Product Details

SNP ID
rs6978233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101314660 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGGAGAGTTCCACGAAGGTGGG[A/T]ATTTCTGTCTTTTTCATTTATGCTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IFT22 PubMed Links

Gene Details

Gene
IFT22
Gene Name
intraflagellar transport 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130820.2 1219 UTR 3 NP_001124292.1
NM_001130821.2 1219 UTR 3 NP_001124293.1
NM_001130822.2 1219 UTR 3 NP_001124294.1
NM_001287525.1 1219 UTR 3 NP_001274454.1
NM_001287526.1 1219 UTR 3 NP_001274455.1
NM_022777.3 1219 UTR 3 NP_073614.1
Gene
LOC101927746
Gene Name
uncharacterized LOC101927746
There are no transcripts associated with this gene.

View Full Product Details