Product Details

SNP ID
rs7320653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:48042061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAAATCCTACCCATAGAGAATT[C/T]AGAGGCTAGGGTTTTTCAAGGATGG
Phenotype
MIM: 615792 MIM: 603921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT15 PubMed Links

Gene Details

Gene
NUDT15
Gene Name
nudix hydrolase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304745.1 Intron NP_001291674.1
NM_018283.3 Intron NP_060753.1
Gene
SUCLA2
Gene Name
succinate-CoA ligase ADP-forming beta subunit
There are no transcripts associated with this gene.

View Full Product Details