Product Details
- SNP ID
-
rs6889921
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:119080887 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGCTTTCTTATTTTCAAGCCATTT[C/T]ACAAGCTCTTCAGGAAACTTTTCAA
- Phenotype
-
MIM: 605671
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DMXL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117929313] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DMXL1
- Gene Name
- Dmx like 1
- Gene
- LOC105379143
- Gene Name
- uncharacterized LOC105379143
There are no transcripts associated with this gene.
View Full Product Details