Product Details

SNP ID
rs7993811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110867684 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGGTGTGGCAGAGATGCAGGGC[C/T]GTGGATCTGATGACACTGCAGCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00346 PubMed Links

Gene Details

Gene
LINC00346
Gene Name
long intergenic non-protein coding RNA 346
There are no transcripts associated with this gene.

View Full Product Details