Product Details

SNP ID
rs8130205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42070063 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCTTGTCATGGCATCTGACATT[C/T]AGTCATTAGCCAAGCAAATGCAGCA
Phenotype
MIM: 613859
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UMODL1 PubMed Links

Gene Details

Gene
UMODL1
Gene Name
uromodulin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004416.2 Intron NP_001004416.2
NM_001199527.1 Intron NP_001186456.1
NM_001199528.2 Intron NP_001186457.2
NM_173568.3 Intron NP_775839.3
XM_011529797.1 Intron XP_011528099.1
XM_017028506.1 Intron XP_016883995.1
XM_017028507.1 Intron XP_016883996.1
XM_017028508.1 Intron XP_016883997.1

View Full Product Details