Product Details

SNP ID
rs7355515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178118878 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTAGAATTCTTAGATACCCCTAA[A/G]GTACATACCCAGATTTTCTCTTAGA
Phenotype
MIM: 608888
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM45 PubMed Links

Gene Details

Gene
RBM45
Gene Name
RNA binding motif protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152945.3 Intron NP_694453.2
XM_005246287.4 Intron XP_005246344.1
XM_017003320.1 Intron XP_016858809.1
XM_017003321.1 Intron XP_016858810.1
XM_017003322.1 Intron XP_016858811.1

View Full Product Details