Product Details

SNP ID
rs7730301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36246963 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTATAATTAGGGTAAACAGTTGG[A/G]TAAAATCTTACTAAAAGAAAGTTAA
Phenotype
MIM: 615787 MIM: 616391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NADK2 PubMed Links

Gene Details

Gene
NADK2
Gene Name
NAD kinase 2, mitochondrial
There are no transcripts associated with this gene.

Gene
RANBP3L
Gene Name
RAN binding protein 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161429.2 4515 UTR 3 NP_001154901.1
NM_001323273.1 4515 UTR 3 NP_001310202.1
NM_001323274.1 4515 Intron NP_001310203.1
NM_001323275.1 4515 UTR 3 NP_001310204.1
NM_001323276.1 4515 Intron NP_001310205.1
NM_001323277.1 4515 Intron NP_001310206.1
NM_001323278.1 4515 UTR 3 NP_001310207.1
NM_001323279.1 4515 UTR 3 NP_001310208.1
NM_001323280.1 4515 Intron NP_001310209.1
NM_145000.4 4515 UTR 3 NP_659437.3
XM_006714452.3 4515 Intron XP_006714515.1
XM_006714453.3 4515 Intron XP_006714516.1
XM_006714455.3 4515 Intron XP_006714518.1

View Full Product Details