Product Details

SNP ID
rs7488249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49022892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGGGGCCTGGGAGGTGATATAA[A/T]CCATGACAAGACAGCTCTCCCTCAG
Phenotype
MIM: 602113 MIM: 602742
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KMT2D PubMed Links

Gene Details

Gene
KMT2D
Gene Name
lysine methyltransferase 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003482.3 Intron NP_003473.3
XM_005269162.4 Intron XP_005269219.1
XM_006719614.3 Intron XP_006719677.1
XM_006719616.3 Intron XP_006719679.1
XM_011538770.2 Intron XP_011537072.1
XM_011538771.2 Intron XP_011537073.1
XM_011538772.2 Intron XP_011537074.1
XM_011538773.2 Intron XP_011537075.1
XM_011538774.2 Intron XP_011537076.1
XM_011538776.2 Intron XP_011537078.1
Gene
PRKAG1
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 1
There are no transcripts associated with this gene.

View Full Product Details