Product Details
- SNP ID
-
hCV29250232
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:50591412 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCAGTGGGGCGGATTTGGGGCA[A/C]TATCATGGCCAGCACACATTAATAG
- Phenotype
-
MIM: 601523
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GRB10
PubMed Links
Gene Details
- Gene
- GRB10
- Gene Name
- growth factor receptor bound protein 10
There are no transcripts associated with this gene.
View Full Product Details