Product Details

SNP ID
rs4463806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:113838234 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCCTTGCTTACTCTAGGACCTAG[C/T]TAATGTAATATTATTTGTTTAATCA
Phenotype
MIM: 609682
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCLRE1A PubMed Links

Gene Details

Gene
DCLRE1A
Gene Name
DNA cross-link repair 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271816.1 Intron NP_001258745.1
NM_014881.4 Intron NP_055696.3
XM_006718090.1 Intron XP_006718153.1
XM_011540429.1 Intron XP_011538731.1

View Full Product Details