Product Details

SNP ID
rs4918066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103860377 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGGCCTCTGGAAGACAAAAAGAA[C/T]AGAAAAGAAAAATACCAGGGGGAAT
Phenotype
MIM: 613128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OBFC1 PubMed Links

Gene Details

Gene
OBFC1
Gene Name
oligonucleotide/oligosaccharide binding fold containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024928.4 Intron NP_079204.2
XM_006717976.3 Intron XP_006718039.1
XM_011540184.2 Intron XP_011538486.1
XM_017016669.1 Intron XP_016872158.1
XM_017016670.1 Intron XP_016872159.1
Gene
SH3PXD2A
Gene Name
SH3 and PX domains 2A
There are no transcripts associated with this gene.

View Full Product Details