Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001077525.2 | Intron | NP_001070993.1 | |||
NM_001077526.2 | Intron | NP_001070994.1 | |||
NM_022485.4 | Intron | NP_071930.2 | |||
XM_005265400.1 | Intron | XP_005265457.1 | |||
XM_011534012.1 | Intron | XP_011532314.1 | |||
XM_011534013.1 | Intron | XP_011532315.1 | |||
XM_017007034.1 | Intron | XP_016862523.1 | |||
XM_017007035.1 | Intron | XP_016862524.1 | |||
XM_017007036.1 | Intron | XP_016862525.1 | |||
XM_017007037.1 | Intron | XP_016862526.1 | |||
XM_017007038.1 | Intron | XP_016862527.1 | |||
XM_017007039.1 | Intron | XP_016862528.1 | |||
XM_017007040.1 | Intron | XP_016862529.1 | |||
XM_017007041.1 | Intron | XP_016862530.1 | |||
XM_017007042.1 | Intron | XP_016862531.1 | |||
XM_017007043.1 | Intron | XP_016862532.1 | |||
XM_017007044.1 | Intron | XP_016862533.1 | |||
XM_017007045.1 | Intron | XP_016862534.1 |