Product Details
- SNP ID
-
rs7643547
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:9917349 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCTGGTTCTTGCTGTCCTTGGCA[C/T]TGGGCCGAAGCCCAGTGGTCCTTTC
- Phenotype
-
MIM: 610925
MIM: 614995
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL17RC
PubMed Links
Gene Details
- Gene
- IL17RC
- Gene Name
- interleukin 17 receptor C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001203263.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
NP_001190192.1 |
NM_001203264.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
NP_001190193.1 |
NM_001203265.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
NP_001190194.1 |
NM_032732.5 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
NP_116121.2 |
NM_153460.3 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
NP_703190.1 |
NM_153461.3 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
NP_703191.1 |
XM_017007326.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862815.1 |
XM_017007327.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862816.1 |
XM_017007328.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862817.1 |
XM_017007329.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862818.1 |
XM_017007330.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862819.1 |
XM_017007331.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862820.1 |
XM_017007332.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862821.1 |
XM_017007333.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862822.1 |
XM_017007334.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862823.1 |
XM_017007335.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862824.1 |
XM_017007336.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862825.1 |
XM_017007337.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862826.1 |
XM_017007338.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862827.1 |
XM_017007339.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862828.1 |
XM_017007340.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862829.1 |
XM_017007341.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862830.1 |
XM_017007342.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862831.1 |
XM_017007343.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862832.1 |
XM_017007344.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862833.1 |
XM_017007345.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862834.1 |
XM_017007346.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862835.1 |
XM_017007347.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862836.1 |
XM_017007348.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862837.1 |
XM_017007349.1 |
276 |
Silent Mutation |
CTG,TTG |
L12L |
XP_016862838.1 |
- Gene
- IL17RE
- Gene Name
- interleukin 17 receptor E
View Full Product Details