Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001112736.1 | 7159 | UTR 3 | NP_001106207.1 | ||
NM_015224.3 | 7159 | UTR 3 | NP_056039.2 | ||
XM_005264999.1 | 7159 | UTR 3 | XP_005265056.1 | ||
XM_006713077.2 | 7159 | Intron | XP_006713140.1 | ||
XM_006713078.2 | 7159 | Intron | XP_006713141.1 | ||
XM_011533552.2 | 7159 | Intron | XP_011531854.1 | ||
XM_011533553.2 | 7159 | Intron | XP_011531855.1 | ||
XM_017006030.1 | 7159 | Intron | XP_016861519.1 | ||
XM_017006031.1 | 7159 | Intron | XP_016861520.1 | ||
XM_017006032.1 | 7159 | Intron | XP_016861521.1 |