Product Details

SNP ID
rs8051433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89697273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACCAGAGGCCCAGACCCCTCCCC[A/G]GCAAAGAGAAGCACCACGGGAGCTG
Phenotype
MIM: 603464
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK10 PubMed Links

Gene Details

Gene
CDK10
Gene Name
cyclin dependent kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098533.2 1840 Intron NP_001092003.2
NM_001160367.1 1840 Intron NP_001153839.1
NM_052987.3 1840 Intron NP_443713.2
NM_052988.4 1840 Intron NP_443714.3
XM_006721308.2 1840 Intron XP_006721371.1
XM_006721310.2 1840 Intron XP_006721373.1
XM_011523405.2 1840 Intron XP_011521707.1
XM_011523406.2 1840 Intron XP_011521708.1
XM_011523407.2 1840 Intron XP_011521709.1
XM_011523408.2 1840 Intron XP_011521710.1
XM_011523410.2 1840 Intron XP_011521712.1
XM_011523411.2 1840 Intron XP_011521713.1
XM_011523412.1 1840 Intron XP_011521714.1
XM_011523413.2 1840 Intron XP_011521715.1
XM_011523414.2 1840 Intron XP_011521716.1
XM_011523415.2 1840 Intron XP_011521717.1
XM_011523416.2 1840 Intron XP_011521718.1
XM_011523417.2 1840 Intron XP_011521719.1
XM_017023806.1 1840 Intron XP_016879295.1
XM_017023807.1 1840 Intron XP_016879296.1
XM_017023808.1 1840 Intron XP_016879297.1
XM_017023809.1 1840 Intron XP_016879298.1
XM_017023810.1 1840 Intron XP_016879299.1
Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152339.3 1840 UTR 3 NP_689552.2
XM_005256279.4 1840 UTR 3 XP_005256336.1
Gene
VPS9D1
Gene Name
VPS9 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details