Product Details
- SNP ID
-
rs7950030
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:18699431 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGGGGGCATTCTCTATGACATCCC[C/G]TGCTTTTCAGACTTTCTCAACTTGA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IGSF22
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75850236] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IGSF22
- Gene Name
- immunoglobulin superfamily member 22
There are no transcripts associated with this gene.
- Gene
- TMEM86A
- Gene Name
- transmembrane protein 86A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153347.2 |
|
Intron |
|
|
NP_699178.1 |
View Full Product Details