Product Details

SNP ID
rs8111600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5821957 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCAAATCCCATGTCCTTTTCTAA[C/T]CCTTGGACAAGCCCCTACCCATTTT
Phenotype
MIM: 136836 MIM: 602018
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUT6 PubMed Links

Gene Details

Gene
FUT6
Gene Name
fucosyltransferase 6
There are no transcripts associated with this gene.

Gene
NRTN
Gene Name
neurturin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011528041.2 Intron XP_011526343.1

View Full Product Details