Product Details

SNP ID
rs8111938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:919983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCGCCCTGGAGCGCGCCTTCGC[A/C]CTGTACAACCTGCTGGCGCTGTACC
Phenotype
MIM: 603265 MIM: 604161
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARID3A PubMed Links

Gene Details

Gene
ARID3A
Gene Name
AT-rich interaction domain 3A
There are no transcripts associated with this gene.

Gene
KISS1R
Gene Name
KISS1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032551.4 776 Silent Mutation GCA,GCC A205A NP_115940.2
XM_017027382.1 776 Intron XP_016882871.1
Gene
R3HDM4
Gene Name
R3H domain containing 4
There are no transcripts associated with this gene.

View Full Product Details