Product Details

SNP ID
rs7558119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218223273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCCCAGGACATGAATCATCTTTT[C/T]GTCTAGCATATCTACAGTGTGTACG
Phenotype
MIM: 604224
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC2 PubMed Links

Gene Details

Gene
ARPC2
Gene Name
actin related protein 2/3 complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005731.3 Intron NP_005722.1
NM_152862.2 Intron NP_690601.1
XM_011510471.1 Intron XP_011508773.1
XM_017003113.1 Intron XP_016858602.1
XM_017003114.1 Intron XP_016858603.1
XM_017003115.1 Intron XP_016858604.1

View Full Product Details