Product Details

SNP ID
rs6661768
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:3736437 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCTCTCAGGCCTCCTGAAGTACA[C/G]GCCCATGAGCCCTCATCCAAGCCCT
Phenotype
MIM: 601990
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TP73 PubMed Links

Gene Details

Gene
TP73
Gene Name
tumor protein p73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126240.2 Intron NP_001119712.1
NM_001126241.2 Intron NP_001119713.1
NM_001126242.2 Intron NP_001119714.1
NM_001204184.1 Intron NP_001191113.1
NM_001204185.1 Intron NP_001191114.1
NM_001204186.1 Intron NP_001191115.1
NM_001204187.1 Intron NP_001191116.1
NM_001204188.1 Intron NP_001191117.1
NM_001204189.1 Intron NP_001191118.1
NM_001204190.1 Intron NP_001191119.1
NM_001204191.1 Intron NP_001191120.1
NM_001204192.1 Intron NP_001191121.1
NM_005427.3 Intron NP_005418.1
Gene
TP73-AS1
Gene Name
TP73 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details