Product Details

SNP ID
rs7127431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67273222 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGGAACCCAGGCACCCGTCCAA[C/T]GGCCAGAGCAAAGGCAACTCTGAGG
Phenotype
MIM: 109635
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRK2 PubMed Links

Gene Details

Gene
GRK2
Gene Name
G protein-coupled receptor kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001619.3 Intron NP_001610.2
XM_011544773.1 Intron XP_011543075.1

View Full Product Details