Product Details
- SNP ID
-
rs7926875
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72138396 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAGTTAGTTTAATAACATGAGAT[A/C]ACTTTTTAAACCGTTAAGAGCTGTA
- Phenotype
-
MIM: 602469
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FOLR3
PubMed Links
Gene Details
- Gene
- FOLR3
- Gene Name
- folate receptor 3
View Full Product Details