Product Details

SNP ID
rs7933235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75829541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGTGAATGCTTGTAAATGCCAG[A/G]GACTGTTTAGGTGCTGGAAGTAAAA
Phenotype
MIM: 602493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UVRAG PubMed Links

Gene Details

Gene
UVRAG
Gene Name
UV radiation resistance associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003369.3 Intron NP_003360.2
XM_011545226.2 Intron XP_011543528.1
XM_011545227.2 Intron XP_011543529.1
XM_011545228.2 Intron XP_011543530.1
XM_017018224.1 Intron XP_016873713.1
XM_017018225.1 Intron XP_016873714.1
XM_017018226.1 Intron XP_016873715.1
XM_017018227.1 Intron XP_016873716.1

View Full Product Details